Assay Details
| Gene Symbol: LINC00708 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_108058.1 | 5 | 667 | 83 | ||
| GenBank mRNA | BC031880.1 | - | 5 | 678 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00708| Gene Symbol: LINC00708 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_108058.1 | 5 | 667 | 83 | ||
| GenBank mRNA | BC031880.1 | - | 5 | 678 | 83 | |
Set Membership: |
|