Assay Details
| Gene Symbol: UTP14C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021645.5 | NP_067677.4 | 2 | 3338 | 113 | |
| GenBank mRNA | BC089407.1 | - | 1 | 2748 | 3065365 | 113 |
| BC122536.1 | - | 2 | 3115 | 40036054 | 113 | |
| D87455.1 | - | 2 | 3338 | 113 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000519151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000519151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000519151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000519151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000519151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000519151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000616513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000616513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NR_036571.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NR_036571.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NR_036571.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NR_036571.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NR_036571.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NR_036571.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000519151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000519151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000519151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000519151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000519151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000519151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000616513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000616513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NR_036571.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NR_036571.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NR_036571.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NR_036571.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NR_036571.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NR_036571.2 | Refseq |

UTP14C
Genomic Map