Assay Details
| Gene Symbol: LINC00641 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038970.1 | 1 | 1233 | 71 | ||
| GenBank mRNA | AK092333.1 | - | 1 | 85 | 71 | |
| BC035145.1 | - | 1 | 1236 | 71 | ||
More Information
Set Membership: |
|

LINC00641
Genomic Map