Assay Details
| Gene Symbol: INF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031714.3 | NP_001026884.3 | 2 | 259 | 74 | |
| NM_022489.3 | NP_071934.3 | 2 | 259 | 74 | ||
| NM_032714.2 | NP_116103.1 | 2 | 259 | 74 | ||
| XM_005268004.4 | XP_005268061.1 | 2 | 346 | 74 | ||
| XM_005268005.4 | XP_005268062.1 | 2 | 317 | 74 | ||
| XM_017021595.1 | XP_016877084.1 | 2 | 317 | 74 | ||
| GenBank mRNA | AK290083.1 | - | 2 | 74 | ||
| BC006173.1 | - | 2 | 74 | |||
| BI489810.1 | - | 2 | 74 | |||
| BX248757.1 | - | 2 | 228 | 74 | ||
More Information
Set Membership: |
|

INF2
Genomic Map