Assay Details
| Gene Symbol: TAGAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278733.1 | NP_001265662.1 | 6 | 1484 | 151 | |
| NM_054114.4 | NP_473455.2 | 10 | 1855 | 151 | ||
| NM_152133.2 | NP_687034.1 | 9 | 1688 | 151 | ||
| GenBank mRNA | AF385429.1 | - | 10 | 1855 | 151 | |
| AF385430.1 | - | 9 | 1688 | 151 | ||
| AK057193.1 | - | 2 | 723 | 151 | ||
| AK097090.1 | - | 10 | 1721 | 151 | ||
| AL832662.1 | - | 10 | 1731 | 151 | ||
| BC111731.1 | - | 10 | 1711 | 40035199 | 151 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |

TAGAP
Genomic Map