Assay Details
| Gene Symbol: LOC105372441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_131203.1 | 3 | 308 | 82 | ||
| NR_131205.1 | 3 | 325 | 82 | |||
| GenBank mRNA | AA565890.1 | - | 1 | 82 | ||
More Information
Set Membership: |
|

LOC105372441
Genomic Map