Assay Details
| Gene Symbol: TMEM161A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256766.1 | NP_001243695.1 | 9 | 897 | 73 | |
| NM_017814.2 | NP_060284.1 | 11 | 1206 | 73 | ||
| XM_011528117.2 | XP_011526419.1 | 6 | 967 | 73 | ||
| XM_017026916.1 | XP_016882405.1 | 11 | 1102 | 73 | ||
| GenBank mRNA | AK000429.1 | - | 11 | 1163 | 73 | |
| AK096964.1 | - | 9 | 862 | 73 | ||
| AK225424.1 | - | 11 | 1120 | 73 | ||
| AY358088.1 | - | 11 | 1190 | 73 | ||
| BC005210.1 | - | 11 | 1166 | 4041226 | 73 | |
| BC016478.2 | - | 10 | 1130 | 73 | ||
More Information
Set Membership: |
|

TMEM161A
Genomic Map