Assay Details
| Gene Symbol: FRMD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC033851.1 | - | 2 | 196 | 102 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000355991.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000355991.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000355991.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000355991.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000355991.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000355991.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000355991.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000355991.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000355991.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000355991.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000355991.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000355991.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000355991.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000416776.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000416776.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000416776.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000416776.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000416776.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000416776.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000416776.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000416776.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000416776.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000416776.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000416776.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000416776.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000525156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000525156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000525156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000526201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000526201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000526201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000526201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000526201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000528854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000528854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000528854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000528854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000528854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000531151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000531151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000531151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000531151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000531151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000531296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000531296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000531296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000533782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000533782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000533782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000533782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000533782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | ENST00000533782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000317568.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000355991.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000355991.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000355991.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000355991.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000355991.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000355991.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000355991.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000355991.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000355991.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000355991.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000355991.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000355991.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000355991.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000416776.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000416776.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000416776.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000416776.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000416776.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000416776.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000416776.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000416776.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000416776.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000416776.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000416776.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000416776.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000525156.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000525156.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000525156.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000526201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000526201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000526201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000526201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000526201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000528854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000528854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000528854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000528854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000528854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000531151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000531151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000531151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000531151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000531151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000531296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000531296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000531296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000533782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000533782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000533782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000533782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000533782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | ENST00000533782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_001300833.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_031904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008015.hg.1 | NM_031904.4 | Refseq |

FRMD8
Genomic Map