Assay Details
| Gene Symbol: HSFY1P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003607.1 | 2 | 1312 | 92 | ||
| GenBank mRNA | AY026053.1 | - | 2 | 1312 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HSFY1P1| Gene Symbol: HSFY1P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003607.1 | 2 | 1312 | 92 | ||
| GenBank mRNA | AY026053.1 | - | 2 | 1312 | 92 | |
Set Membership: |
|