Assay Details
| Gene Symbol: D2HGDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF318334.1 | - | 2 | 986 | 90 | |
| AK055846.1 | - | 3 | 1002 | 90 | ||
| AK096060.1 | - | 2 | 916 | 90 | ||
| AK124189.1 | - | 2 | 2595 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK055846.1, AK096060.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000321264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000321264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000321264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000321264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000321264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000321264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000321264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000321264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000321264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000321264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000321264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000321264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000321264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000400769.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000400769.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000400769.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000400769.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000400769.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000400769.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000400769.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000400769.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000400769.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000400769.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000400769.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000403782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000403782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000403782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000403782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000403782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000403782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000403782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000403782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000403782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000403782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000403782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000417686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000417686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000417686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000417686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000432449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000432449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000432449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000432449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000432449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000432449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000436747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000436747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000436747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000436747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000436747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000436747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000436747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000436747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000436747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000436747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000436747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000436747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000436747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000436747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000436747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000436747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000436747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000436747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000436747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000436747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000436747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000436747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000436747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000436747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000436747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000437164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000437164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000437164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000445308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000445308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000445308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000445308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000454048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000454048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000463932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000463932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000463932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000467427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000467427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000467427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000467427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000467427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000467427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000467427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000468064.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000468064.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000468064.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000468064.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000468064.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000468064.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000468064.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000468064.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000468064.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000470343.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000470343.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000470343.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000470343.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000470343.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000470343.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000470343.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000473126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000473126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000473126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000473126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000473126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000473126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000486953.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000486953.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000486953.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000486953.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000486953.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000486953.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000486953.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000486953.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000496252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000496252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000496252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000496252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000496252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000496252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000610344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000610344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000610344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000610344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000610344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000610344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000610344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000610344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | ENST00000610344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NM_001287249.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NM_001287249.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NM_001287249.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NM_001287249.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NM_001287249.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NM_001287249.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NM_001287249.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NM_001287249.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NM_001287249.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NM_001287249.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NM_001287249.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NM_152783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NM_152783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NM_152783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NM_152783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NM_152783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NM_152783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NM_152783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NM_152783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NM_152783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NM_152783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NM_152783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NM_152783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NM_152783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NR_109778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NR_109778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NR_109778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NR_109778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NR_109778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NR_109778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NR_109778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NR_109778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NR_109778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NR_109778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011423.hg.1 | NR_109778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016376.hg.1 | ENST00000417267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016376.hg.1 | ENST00000417267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016376.hg.1 | ENST00000417267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016376.hg.1 | ENST00000417267.1 | Ensembl |

D2HGDH
Genomic Map