Assay Details
Gene Symbol: ZNRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032268.4 | NP_115644.1 | 5 | 3284 | 107 | |
XM_017023793.1 | XP_016879282.1 | 5 | 1589 | 107 | ||
GenBank mRNA | AK027752.1 | - | 1 | 1045 | 107 | |
AL390144.1 | - | 1 | 4263 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600010865.hg.1 | ENST00000499110.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010865.hg.1 | ENST00000499110.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000320619.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000320619.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000320619.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000320619.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000320619.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000320619.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000320619.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000320619.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000320619.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000320619.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000320619.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000320619.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000320619.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000564320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000564320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000564320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000564320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000564320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000564320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000564320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000566244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000566244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000566244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000566244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000566244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000566244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000566244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000566244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000566244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000566244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000566244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000566244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000566250.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000566250.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000566250.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000566250.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000566250.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000567962.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000567962.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000567962.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000567962.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000567962.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000567962.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000567962.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568511.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568511.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568511.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568511.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000568844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000579084.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000579084.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | ENST00000579084.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000320619.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000320619.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000320619.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000320619.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000320619.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000320619.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000320619.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000320619.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000320619.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000320619.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000320619.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000320619.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000320619.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000335325.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000564320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000564320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000564320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000564320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000564320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000564320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000564320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000566244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000566244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000566244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000566244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000566244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000566244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000566244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000566244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000566244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000566244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000566244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000566244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000566250.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000566250.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000566250.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000566250.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000566250.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000567962.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000567962.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000567962.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000567962.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000567962.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000567962.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000567962.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568511.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568511.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568511.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568511.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000568844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000579084.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000579084.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | ENST00000579084.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008427.hg.1 | NM_032268.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010865.hg.1 | ENST00000499110.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010865.hg.1 | ENST00000499110.1 | Ensembl |
