Assay Details
| Gene Symbol: HERC2P4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC038215.2 | - | 2 | 520 | 77 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000564458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000564458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000564458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000564145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000564145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000564145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000564145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000564145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000570151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000570151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011530.hg.1 | ENST00000564165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011530.hg.1 | ENST00000564165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011530.hg.1 | ENST00000564165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011530.hg.1 | ENST00000564165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011530.hg.1 | ENST00000564165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011530.hg.1 | ENST00000564795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011530.hg.1 | ENST00000564795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011530.hg.1 | ENST00000564795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000564458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000564458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000564458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000564145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000564145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000564145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000564145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000564145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000570151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000570151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011530.hg.1 | ENST00000564165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011530.hg.1 | ENST00000564165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011530.hg.1 | ENST00000564165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011530.hg.1 | ENST00000564165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011530.hg.1 | ENST00000564165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011530.hg.1 | ENST00000564795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011530.hg.1 | ENST00000564795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011530.hg.1 | ENST00000564795.1 | Ensembl |

HERC2P4
Genomic Map