Assay Details
| Gene Symbol: LINC01106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027244.1 | 2 | 1300 | 88 | ||
| GenBank mRNA | AK095678.1 | - | 2 | 1610 | 88 | |
| CR749351.1 | - | 3 | 1393 | 88 | ||
More Information
Set Membership: |
|

LINC01106
Genomic Map