Assay Details
| Gene Symbol: LOC101929054 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135307.1 | 2 | 879 | 110 | ||
| GenBank mRNA | BC039681.1 | - | 2 | 853 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929054| Gene Symbol: LOC101929054 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135307.1 | 2 | 879 | 110 | ||
| GenBank mRNA | BC039681.1 | - | 2 | 853 | 110 | |
Set Membership: |
|