Assay Details
| Gene Symbol: ZNF445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK310588.1 | - | 2 | 804 | 75 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000474600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000474600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000474600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000474600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000474600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000474600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000474600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000474600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000474600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000474600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |

ZNF445
Genomic Map