Assay Details
| Gene Symbol: SMG1P5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CR627362.1 | - | 2 | 3818 | 69 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000532983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000532983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000532983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000532983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000532983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000532983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000532983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000532983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000532983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000532983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000532983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000532983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | ENST00000620872.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007202.hg.1 | ENST00000610337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015254.hg.1 | ENST00000617525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011503.hg.1 | ENST00000616299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009924.hg.1 | ENST00000612780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016105.hg.1 | ENST00000615649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011514.hg.1 | ENST00000621224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011506.hg.1 | ENST00000617496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016105.hg.1 | ENST00000615649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007202.hg.1 | ENST00000610337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011506.hg.1 | ENST00000617496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011514.hg.1 | ENST00000621224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015254.hg.1 | ENST00000617525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000411546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000529428.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000532983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000532983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000532983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000532983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000532983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000532983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000532983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000532983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000532983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000532983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000532983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000532983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | ENST00000620872.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009959.hg.1 | NR_002453.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009924.hg.1 | ENST00000612780.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011503.hg.1 | ENST00000616299.1 | Ensembl |

SMG1P5
Genomic Map