Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013707.hg.1 |
ENST00000332629.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013707.hg.1 |
ENST00000332629.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013707.hg.1 |
ENST00000332629.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013707.hg.1 |
ENST00000332629.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013707.hg.1 |
ENST00000332629.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013707.hg.1 |
ENST00000332629.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013707.hg.1 |
ENST00000425888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013707.hg.1 |
ENST00000425888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013707.hg.1 |
ENST00000425888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013707.hg.1 |
ENST00000425888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013707.hg.1 |
ENST00000429115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013707.hg.1 |
ENST00000429115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013707.hg.1 |
ENST00000433160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013707.hg.1 |
ENST00000433160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013707.hg.1 |
ENST00000433160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013707.hg.1 |
ENST00000433160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013707.hg.1 |
ENST00000433160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013707.hg.1 |
ENST00000456677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013707.hg.1 |
ENST00000456677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013707.hg.1 |
NM_182627.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013707.hg.1 |
NM_182627.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013707.hg.1 |
NM_182627.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013707.hg.1 |
NM_182627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013707.hg.1 |
ENST00000332629.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013707.hg.1 |
ENST00000332629.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013707.hg.1 |
ENST00000332629.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013707.hg.1 |
ENST00000332629.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013707.hg.1 |
ENST00000332629.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013707.hg.1 |
ENST00000332629.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013707.hg.1 |
ENST00000425888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013707.hg.1 |
ENST00000425888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013707.hg.1 |
ENST00000425888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013707.hg.1 |
ENST00000425888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013707.hg.1 |
ENST00000429115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013707.hg.1 |
ENST00000429115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013707.hg.1 |
ENST00000433160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013707.hg.1 |
ENST00000433160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013707.hg.1 |
ENST00000433160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013707.hg.1 |
ENST00000433160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013707.hg.1 |
ENST00000433160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013707.hg.1 |
ENST00000456677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013707.hg.1 |
ENST00000456677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013707.hg.1 |
NM_182627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013707.hg.1 |
NM_182627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013707.hg.1 |
NM_182627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013707.hg.1 |
NM_182627.2 |
Refseq |