Assay Details
| Gene Symbol: TBCCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134415.1 | NP_001127887.1 | 2 | 578 | 65 | |
| NM_018138.4 | NP_060608.1 | 2 | 229 | 65 | ||
| GenBank mRNA | AK001422.1 | - | 2 | 202 | 65 | |
| BC025748.1 | - | 2 | 215 | 5200082 | 65 | |
| DB023715.1 | - | 2 | 227 | 65 | ||
| HY017180.1 | - | 2 | 229 | 65 | ||
More Information
Set Membership: |
|

TBCCD1
Genomic Map