| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000366506.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000366506.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000366506.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000366506.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000366506.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000366506.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |