Assay Details
| Gene Symbol: TLR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK302143.1 | - | 1 | 126 | 81 | |
| BC043204.2 | - | 1 | 132 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK302143.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000296795.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000296795.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000296795.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000296795.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000296795.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000296795.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000296795.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000296795.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000296795.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000296795.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000296795.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000296795.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000504367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000504367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000504367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000504367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000504367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000504367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000504367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000508051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000508051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000508051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000512264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000512264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000512264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000512264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000512264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000512264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000512264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000512264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000512264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000513189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000513189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000513189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000513189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | ENST00000513189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | NM_003265.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | NM_003265.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | NM_003265.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | NM_003265.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | NM_003265.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | NM_003265.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | NM_003265.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | NM_003265.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | NM_003265.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | NM_003265.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009543.hg.1 | NM_003265.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000296795.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000296795.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000296795.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000296795.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000296795.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000296795.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000296795.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000296795.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000296795.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000296795.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000296795.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000296795.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000504367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000504367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000504367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000504367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000504367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000504367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000504367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000508051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000508051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000508051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000512264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000512264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000512264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000512264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000512264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000512264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000512264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000512264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000512264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000513189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000513189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000513189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000513189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | ENST00000513189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | NM_003265.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | NM_003265.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | NM_003265.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | NM_003265.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | NM_003265.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | NM_003265.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | NM_003265.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | NM_003265.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | NM_003265.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | NM_003265.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009543.hg.1 | NM_003265.2 | Refseq |

TLR3
Genomic Map