Assay Details
| Gene Symbol: APELA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017007623.1 | XP_016863112.1 | 3 | 550 | 110 | |
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000505269.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000505269.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000505269.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000505269.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000510062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000510062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000510062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000510062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000510062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000510062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000510062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000510062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000515275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000515275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000515275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000515275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000515405.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000515405.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | ENST00000515405.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | NM_001297550.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | NM_001297550.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | NM_001297550.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | NM_001297550.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | NM_001297550.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | NM_001297550.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | NM_001297550.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | NM_001297550.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | NM_001297550.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | NM_001297550.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | NM_001297550.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009198.hg.1 | NM_001297550.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000505269.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000505269.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000505269.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000505269.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000507152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000510062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000510062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000510062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000510062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000510062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000510062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000510062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000510062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000515275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000515275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000515275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000515275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000515405.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000515405.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | ENST00000515405.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | NM_001297550.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | NM_001297550.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | NM_001297550.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | NM_001297550.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | NM_001297550.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | NM_001297550.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | NM_001297550.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | NM_001297550.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | NM_001297550.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | NM_001297550.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | NM_001297550.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009198.hg.1 | NM_001297550.1 | Refseq |

APELA
Genomic Map