| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010082.hg.1 |
ENST00000627163.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010082.hg.1 |
ENST00000627163.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010082.hg.1 |
ENST00000627163.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010082.hg.1 |
ENST00000627163.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010082.hg.1 |
ENST00000627163.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010082.hg.1 |
ENST00000627163.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010082.hg.1 |
ENST00000627163.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010082.hg.1 |
ENST00000627575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010082.hg.1 |
ENST00000627575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010082.hg.1 |
ENST00000627575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010082.hg.1 |
ENST00000627575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010082.hg.1 |
ENST00000627575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010082.hg.1 |
ENST00000627575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010082.hg.1 |
ENST00000628853.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010082.hg.1 |
ENST00000628853.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010082.hg.1 |
ENST00000628853.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010082.hg.1 |
NR_034054.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010082.hg.1 |
NR_034054.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010082.hg.1 |
NR_034054.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010082.hg.1 |
NR_034054.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010082.hg.1 |
NR_034054.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010082.hg.1 |
NR_034054.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010082.hg.1 |
NR_034054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010082.hg.1 |
ENST00000627163.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010082.hg.1 |
ENST00000627163.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010082.hg.1 |
ENST00000627163.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010082.hg.1 |
ENST00000627163.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010082.hg.1 |
ENST00000627163.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010082.hg.1 |
ENST00000627163.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010082.hg.1 |
ENST00000627163.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010082.hg.1 |
ENST00000627575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010082.hg.1 |
ENST00000627575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010082.hg.1 |
ENST00000627575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010082.hg.1 |
ENST00000627575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010082.hg.1 |
ENST00000627575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010082.hg.1 |
ENST00000627575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010082.hg.1 |
ENST00000628853.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010082.hg.1 |
ENST00000628853.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010082.hg.1 |
ENST00000628853.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010082.hg.1 |
NR_034054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010082.hg.1 |
NR_034054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010082.hg.1 |
NR_034054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010082.hg.1 |
NR_034054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010082.hg.1 |
NR_034054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010082.hg.1 |
NR_034054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010082.hg.1 |
NR_034054.1 |
Refseq |