Assay Details
| Gene Symbol: AGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000027.3 | NP_000018.2 | 9 | 1593 | 67 | |
| NM_001171988.1 | NP_001165459.1 | 9 | 1563 | 67 | ||
| NR_033655.1 | 8 | 1517 | 67 | |||
| GenBank mRNA | BM968469.1 | - | 1 | 67 | ||
More Information
Set Membership: |
|

AGA
Genomic Map