Assay Details
Gene Symbol: LINC01088 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038342.1 | 6 | 869 | 110 | ||
GenBank mRNA | AA195044.1 | - | 1 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000437737.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000437737.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000505149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000505149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000505149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000505149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000505149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000505149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000505149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000505149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000507761.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000507761.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000507761.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000510667.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000510667.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000510667.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000510667.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000510667.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000510667.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000510667.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000510667.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000511241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000511241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000511241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000511241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000511241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000511241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000511241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000511241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000511241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000511241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000511241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000511241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000511895.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000511895.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000512130.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000512130.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000512130.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000512130.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000512130.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000512130.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000512130.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000512130.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000512130.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000512130.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000512130.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000512130.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000515597.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | ENST00000515597.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | NR_038342.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | NR_038342.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | NR_038342.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | NR_038342.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | NR_038342.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | NR_038342.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | NR_038342.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | NR_038342.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | NR_038342.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | NR_038342.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007943.hg.1 | NR_038342.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000437737.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000437737.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000505149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000505149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000505149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000505149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000505149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000505149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000505149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000505149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000507476.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000507761.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000507761.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000507761.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000509088.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000510667.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000510667.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000510667.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000510667.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000510667.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000510667.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000510667.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000510667.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000511241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000511241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000511241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000511241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000511241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000511241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000511241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000511241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000511241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000511241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000511241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000511241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000511895.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000511895.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000512130.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000512130.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000512130.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000512130.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000512130.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000512130.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000512130.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000512130.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000512130.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000512130.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000512130.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000512130.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000513153.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000515597.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | ENST00000515597.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | NR_038342.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | NR_038342.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | NR_038342.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | NR_038342.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | NR_038342.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | NR_038342.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | NR_038342.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | NR_038342.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | NR_038342.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | NR_038342.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007943.hg.1 | NR_038342.1 | Refseq |
