Assay Details
| Gene Symbol: RGS7BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029875.2 | NP_001025046.1 | 1 | 430 | 99 | |
| XM_005248502.3 | XP_005248559.1 | 1 | 1812 | 99 | ||
| GenBank mRNA | AK296443.1 | - | 1 | 78 | 99 | |
| DA404068.1 | - | 1 | 430 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK296443.1, NM_001029875.2, XM_005248502.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000508162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000508162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000508162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000508162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000508162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000508162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000508162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000508162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NR_073511.1 | Refseq |

RGS7BP
Genomic Map