Assay Details
| Gene Symbol: TP53AIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251964.1 | NP_001238893.1 | 2 | 1580 | 64 | |
| GenBank mRNA | AB045832.1 | - | 2 | 1108 | 64 | |
| AK310612.1 | - | 2 | 1266 | 64 | ||
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000458238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000458238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000458238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000458238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000458238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000458238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000527504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000527504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000527504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000527504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000527504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000527504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000527504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000527504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000530777.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000530777.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000530777.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000530777.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000531399.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000531399.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000531399.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000531399.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000531399.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000531399.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000531399.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000621818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000621818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000621818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000621818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000621818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000621818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | ENST00000621818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001195194.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001195194.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001195194.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001195194.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001195194.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001195195.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001195195.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001195195.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001195195.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001195195.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001195195.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001195195.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001195195.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_022112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_022112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_022112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_022112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_022112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_022112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_022112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_022112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012793.hg.1 | NM_022112.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000458238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000458238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000458238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000458238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000458238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000458238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000525390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000527504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000527504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000527504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000527504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000527504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000527504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000527504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000527504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000530777.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000530777.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000530777.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000530777.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000531399.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000531399.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000531399.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000531399.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000531399.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000531399.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000531399.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000602346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000621818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000621818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000621818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000621818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000621818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000621818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | ENST00000621818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001195194.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001195194.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001195194.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001195194.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001195194.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001195195.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001195195.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001195195.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001195195.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001195195.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001195195.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001195195.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001195195.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_001251964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_022112.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_022112.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_022112.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_022112.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_022112.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_022112.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_022112.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_022112.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012793.hg.1 | NM_022112.2 | Refseq |

TP53AIP1
Genomic Map