| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007396.hg.1 |
ENST00000481727.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007396.hg.1 |
ENST00000481727.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007396.hg.1 |
ENST00000481727.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007396.hg.1 |
ENST00000481727.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007396.hg.1 |
ENST00000491143.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007396.hg.1 |
ENST00000491143.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007396.hg.1 |
ENST00000491143.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007396.hg.1 |
ENST00000491143.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007396.hg.1 |
ENST00000491143.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007396.hg.1 |
ENST00000491143.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007396.hg.1 |
ENST00000491143.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007396.hg.1 |
ENST00000491143.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007396.hg.1 |
ENST00000581316.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007396.hg.1 |
NM_004852.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007396.hg.1 |
NM_004852.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007396.hg.1 |
NM_004852.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007396.hg.1 |
NM_004852.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007396.hg.1 |
NM_004852.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007396.hg.1 |
NM_004852.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007396.hg.1 |
NM_004852.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007396.hg.1 |
NM_004852.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008897.hg.1 |
ENST00000328345.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008897.hg.1 |
ENST00000328345.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008897.hg.1 |
ENST00000328345.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008897.hg.1 |
ENST00000328345.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008897.hg.1 |
ENST00000328345.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008897.hg.1 |
ENST00000328345.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008897.hg.1 |
NM_005604.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008897.hg.1 |
NM_005604.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008897.hg.1 |
NM_005604.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008897.hg.1 |
NM_005604.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008897.hg.1 |
ENST00000328345.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008897.hg.1 |
ENST00000328345.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008897.hg.1 |
ENST00000328345.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008897.hg.1 |
ENST00000328345.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008897.hg.1 |
ENST00000328345.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008897.hg.1 |
ENST00000328345.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008897.hg.1 |
NM_005604.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008897.hg.1 |
NM_005604.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008897.hg.1 |
NM_005604.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008897.hg.1 |
NM_005604.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007396.hg.1 |
ENST00000481727.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007396.hg.1 |
ENST00000481727.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007396.hg.1 |
ENST00000481727.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007396.hg.1 |
ENST00000481727.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007396.hg.1 |
ENST00000491143.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007396.hg.1 |
ENST00000491143.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007396.hg.1 |
ENST00000491143.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007396.hg.1 |
ENST00000491143.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007396.hg.1 |
ENST00000491143.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007396.hg.1 |
ENST00000491143.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007396.hg.1 |
ENST00000491143.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007396.hg.1 |
ENST00000491143.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007396.hg.1 |
ENST00000581316.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007396.hg.1 |
NM_004852.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007396.hg.1 |
NM_004852.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007396.hg.1 |
NM_004852.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007396.hg.1 |
NM_004852.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007396.hg.1 |
NM_004852.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007396.hg.1 |
NM_004852.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007396.hg.1 |
NM_004852.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007396.hg.1 |
NM_004852.2 |
Refseq |