Assay Details
| Gene Symbol: STXBP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127715.2 | NP_001121187.1 | 28 | 8987 | 80 | |
| NM_139244.4 | NP_640337.3 | 26 | 8879 | 80 | ||
| XM_006715339.2 | XP_006715402.1 | 27 | 9113 | 80 | ||
| XM_011535453.1 | XP_011533755.1 | 27 | 8711 | 80 | ||
| XM_017010267.1 | XP_016865756.1 | 23 | 9058 | 80 | ||
| GenBank mRNA | BC043173.2 | - | 2 | 1123 | 80 | |
More Information
Set Membership: |
|

STXBP5
Genomic Map