Assay Details
| Gene Symbol: TTBK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017011367.1 | XP_016866856.1 | 12 | 2678 | 153 | |
| GenBank mRNA | AK131217.1 | - | 14 | 2217 | 153 | |
More Information
Set Membership: |
|

TTBK1
Genomic Map