Assay Details
| Gene Symbol: LOC100507194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038295.1 | 3 | 732 | 94 | ||
| NR_038296.1 | 3 | 705 | 94 | |||
| GenBank mRNA | AL528822.3 | - | 3 | 704 | 94 | |
| BC027024.1 | - | 3 | 722 | 94 | ||
More Information
Set Membership: |
|

LOC100507194
Genomic Map