Assay Details
| Gene Symbol: TNXB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019105.6 | NP_061978.6 | 2 | 374 | 88 | |
| GenBank mRNA | AK310080.1 | - | 2 | 313 | 88 | |
| X71923.1 | - | 2 | 220 | 88 | ||
More Information
Set Membership: |
|

TNXB
Genomic Map