Assay Details
| Gene Symbol: PMS2P5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027777.2 | 4 | 732 | 104 | ||
| GenBank mRNA | BC052588.1 | - | 4 | 735 | 104 | |
| BX116204.1 | - | 1 | 104 | |||
| Gene Symbol: PMS2P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC110395.1 | - | 6 | 921 | 104 | |
More Information
Set Membership: |
|

PMS2P5
Genomic Map