Assay Details
| Gene Symbol: LOC100507642 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_108064.1 | 3 | 2042 | 90 | ||
| NR_108065.1 | 2 | 1935 | 90 | |||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000471299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000471299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000471299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000471299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000479592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000479592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000479592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000479592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000492208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000492208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000492208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000471299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000471299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000471299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000471299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000479592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000479592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000479592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000479592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000492208.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000492208.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000492208.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | NR_108064.1 | Refseq |

LOC100507642
Genomic Map