Assay Details
| Gene Symbol: HOTAIRM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038366.1 | 1 | 121 | 94 | ||
| NR_038367.1 | 1 | 121 | 94 | |||
| GenBank mRNA | BC031342.1 | - | 1 | 91 | 94 | |
| GQ479958.1 | - | 1 | 121 | 94 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000425358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000425358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000425358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000425358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000425358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000428939.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000428939.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000428939.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000428939.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000428939.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000428939.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000428939.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000434063.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000434063.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000434063.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000434063.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000434063.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000434063.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000434063.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000434063.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000434063.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000495032.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000495032.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000495032.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000593300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000593300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000593300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000593300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000593300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000593300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000616712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000617934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000619311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000619974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000622675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | ENST00000622675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | NR_038366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | NR_038366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | NR_038366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | NR_038366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | NR_038366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | NR_038366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | NR_038366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | NR_038366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | NR_038366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | NR_038366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | NR_038366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | NR_038367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | NR_038367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | NR_038367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | NR_038367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | NR_038367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | NR_038367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | NR_038367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | NR_038367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | NR_038367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007000.hg.1 | NR_038367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000425358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000425358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000425358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000425358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000425358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000428939.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000428939.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000428939.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000428939.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000428939.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000428939.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000428939.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000429611.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000434063.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000434063.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000434063.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000434063.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000434063.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000434063.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000434063.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000434063.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000434063.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000495032.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000495032.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000495032.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000593300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000593300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000593300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000593300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000593300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000593300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000616712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000617934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000619311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000619974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000622675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | ENST00000622675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | NR_038366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | NR_038366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | NR_038366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | NR_038366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | NR_038366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | NR_038366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | NR_038366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | NR_038366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | NR_038366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | NR_038366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | NR_038366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | NR_038367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | NR_038367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | NR_038367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | NR_038367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | NR_038367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | NR_038367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | NR_038367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | NR_038367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | NR_038367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007000.hg.1 | NR_038367.1 | Refseq |

HOTAIRM1
Genomic Map