| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009646.hg.1 |
ENST00000415124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009646.hg.1 |
ENST00000415124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009646.hg.1 |
ENST00000415124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009646.hg.1 |
ENST00000415124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009646.hg.1 |
ENST00000417897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009646.hg.1 |
ENST00000417897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009646.hg.1 |
ENST00000417897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009646.hg.1 |
ENST00000572046.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009646.hg.1 |
ENST00000572046.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009646.hg.1 |
ENST00000572046.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009646.hg.1 |
ENST00000573187.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009646.hg.1 |
ENST00000573187.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009646.hg.1 |
ENST00000573187.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009646.hg.1 |
ENST00000573187.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009646.hg.1 |
ENST00000415124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009646.hg.1 |
ENST00000415124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009646.hg.1 |
ENST00000415124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009646.hg.1 |
ENST00000415124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009646.hg.1 |
ENST00000417897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009646.hg.1 |
ENST00000417897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009646.hg.1 |
ENST00000417897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009646.hg.1 |
ENST00000572046.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009646.hg.1 |
ENST00000572046.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009646.hg.1 |
ENST00000572046.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009646.hg.1 |
ENST00000573187.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009646.hg.1 |
ENST00000573187.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009646.hg.1 |
ENST00000573187.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009646.hg.1 |
ENST00000573187.1 |
Ensembl |