Assay Details
| Gene Symbol: HRAT17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110162.1 | 5 | 97 | |||
| GenBank mRNA | AA707363.1 | - | 2 | 228 | 97 | |
| AA992588.1 | - | 2 | 220 | 97 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HRAT17| Gene Symbol: HRAT17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110162.1 | 5 | 97 | |||
| GenBank mRNA | AA707363.1 | - | 2 | 228 | 97 | |
| AA992588.1 | - | 2 | 220 | 97 | ||
Set Membership: |
|