Assay Details
| Gene Symbol: KIF13B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015254.3 | NP_056069.2 | 40 | 8641 | 142 | |
| XM_005273458.1 | XP_005273515.1 | 39 | 8578 | 142 | ||
| XM_011544457.1 | XP_011542759.1 | 40 | 8704 | 142 | ||
| XM_011544458.1 | XP_011542760.1 | 39 | 8641 | 142 | ||
| XM_011544459.2 | XP_011542761.1 | 40 | 8689 | 142 | ||
| XM_011544460.2 | XP_011542762.1 | 40 | 8696 | 142 | ||
| XM_011544461.1 | XP_011542763.1 | 39 | 8691 | 142 | ||
| GenBank mRNA | AB014539.3 | - | 40 | 8611 | 142 | |
| AF279865.1 | - | 40 | 8619 | 142 | ||
| AI760125.1 | - | 1 | 142 | |||
| AL583912.1 | - | 14 | 5299 | 142 | ||
More Information
Set Membership: |
|

KIF13B
Genomic Map