Assay Details
| Gene Symbol: FZD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017412.3 | NP_059108.1 | 3 | 277 | 95 | |
| NM_145866.1 | NP_665873.1 | 2 | 231 | 95 | ||
| XM_017013842.1 | XP_016869331.1 | 3 | 267 | 95 | ||
| XM_017013843.1 | XP_016869332.1 | 3 | 267 | 95 | ||
| XM_017013844.1 | XP_016869333.1 | 3 | 267 | 95 | ||
| GenBank mRNA | AB039723.1 | - | 3 | 228 | 95 | |
| AJ272427.1 | - | 1 | 84 | 95 | ||
| AK291480.1 | - | 2 | 226 | 95 | ||
| BC034278.1 | - | 1 | 76 | 95 | ||
| DA250029.1 | - | 3 | 274 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB039723.1, AJ272427.1, AK291480.1, NM_017412.3, NM_145866.1, XM_017013842.1, XM_017013843.1, XM_017013844.1 transcripts.
Set Membership: |
|

FZD3
Genomic Map