Assay Details
| Gene Symbol: ZNF252P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_023392.1 | 5 | 5321 | 90 | ||
| GenBank mRNA | AK128723.1 | - | 1 | 3646 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF252P| Gene Symbol: ZNF252P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_023392.1 | 5 | 5321 | 90 | ||
| GenBank mRNA | AK128723.1 | - | 1 | 3646 | 90 | |
Set Membership: |
|