Assay Details
| Gene Symbol: AQP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK095363.1 | - | 3 | 1783 | 92 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000494313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000494313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000494313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000494313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000494313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000494313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000494313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000494313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000494313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000494313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000494313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000494313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | NM_004925.4 | Refseq |

AQP3
Genomic Map