Assay Details
| Gene Symbol: LOC105376037 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK098263.1 | - | 2 | 1320 | 84 | |
| AK126527.1 | - | 1 | 2668 | 84 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105376037| Gene Symbol: LOC105376037 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK098263.1 | - | 2 | 1320 | 84 | |
| AK126527.1 | - | 1 | 2668 | 84 | ||
Set Membership: |
|