Assay Details
| Gene Symbol: TRAPPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011658.3 | NP_001011658.1 | 6 | 2796 | 109 | |
| NM_001128835.2 | NP_001122307.2 | 6 | 2790 | 109 | ||
| NM_014563.5 | NP_055378.1 | 5 | 2654 | 109 | ||
| XM_011545565.1 | XP_011543867.1 | 7 | 2864 | 109 | ||
| XM_011545566.2 | XP_011543868.1 | 6 | 2720 | 109 | ||
| GenBank mRNA | AA770629.1 | - | 1 | 74 | 109 | |
| AK055671.1 | - | 1 | 1961 | 109 | ||
| BC016915.1 | - | 6 | 1920 | 4432196 | 109 | |
| BC052618.1 | - | 5 | 2627 | 6581382 | 109 | |
| BM931610.1 | - | 1 | 468 | 109 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000517553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000517553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000517553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000517553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000517553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000518847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000518847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000518847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000518847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000518847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000518847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000519885.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000519885.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000519885.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000519885.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000517553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000517553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000517553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000517553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000517553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000518847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000518847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000518847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000518847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000518847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000518847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000519885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000519885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000519885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000519885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_014563.5 | Refseq |

TRAPPC2
Genomic Map