Assay Details
| Gene Symbol: OAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320151.1 | NP_001307080.1 | 6 | 134 | ||
| GenBank mRNA | AA085948.1 | - | 1 | 217 | 133 | |
| AJ629455.1 | - | 6 | 133 | |||
| DQ823040.1 | - | 2 | 134 | |||
| DQ914956.1 | - | 2 | 133 | |||
More Information
Set Membership: |
|

OAS1
Genomic Map