Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000445759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000445759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000445759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000445759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000446986.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000446986.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000446986.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000446986.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000609227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000609227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000445759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000445759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000445759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000445759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000446986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000446986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000446986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000446986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000609227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000609227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |