Assay Details
| Gene Symbol: RAX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013435.2 | NP_038463.2 | 2 | 522 | 92 | |
| GenBank mRNA | AF115392.1 | - | 2 | 525 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RAX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013435.2 | NP_038463.2 | 2 | 522 | 92 | |
| GenBank mRNA | AF115392.1 | - | 2 | 525 | 92 | |
Set Membership: |
|