Assay Details
| Gene Symbol: MLEC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001303627.1 | NP_001290556.1 | 5 | 3448 | 71 | |
| NM_001303628.1 | NP_001290557.1 | 3 | 3496 | 71 | ||
| NM_014730.3 | NP_055545.1 | 5 | 3731 | 71 | ||
| XM_011539031.1 | XP_011537333.1 | 5 | 8146 | 71 | ||
| XM_011539032.1 | XP_011537334.1 | 6 | 7834 | 71 | ||
| GenBank mRNA | CR936818.1 | - | 2 | 2900 | 71 | |
| D63486.1 | - | 5 | 3688 | 71 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200012176.hg.1 | ENST00000535720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012176.hg.1 | ENST00000535720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012176.hg.1 | ENST00000535720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012176.hg.1 | ENST00000535720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012176.hg.1 | ENST00000541383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012176.hg.1 | ENST00000541383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000535413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000535413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000535656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000535656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000535656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000535656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000535656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000535656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000545525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000545525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000545525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000545525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000545525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000545525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000535413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000535413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000535656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000535656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000535656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000535656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000535656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000535656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000545525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000545525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000545525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000545525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000545525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000545525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012176.hg.1 | ENST00000535720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012176.hg.1 | ENST00000535720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012176.hg.1 | ENST00000535720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012176.hg.1 | ENST00000535720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012176.hg.1 | ENST00000541383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012176.hg.1 | ENST00000541383.1 | Ensembl |

MLEC
Genomic Map