Assay Details
| Gene Symbol: LINC00592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027358.1 | 3 | 325 | 67 | ||
| GenBank mRNA | BC039104.1 | - | 3 | 325 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00592| Gene Symbol: LINC00592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027358.1 | 3 | 325 | 67 | ||
| GenBank mRNA | BC039104.1 | - | 3 | 325 | 67 | |
Set Membership: |
|