Assay Details
| Gene Symbol: XLOC_009911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_131215.1 | 2 | 371 | 106 | ||
| GenBank mRNA | HY014193.1 | - | 2 | 372 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

XLOC_009911| Gene Symbol: XLOC_009911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_131215.1 | 2 | 371 | 106 | ||
| GenBank mRNA | HY014193.1 | - | 2 | 372 | 106 | |
Set Membership: |
|