Assay Details
| Gene Symbol: LOC283352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104172.1 | 2 | 124 | 119 | ||
| NR_104173.1 | 2 | 124 | 119 | |||
| GenBank mRNA | AK097496.1 | - | 1 | 1372 | 119 | |
| BC046203.1 | - | 2 | 120 | 118 | ||
| BM823446.1 | - | 2 | 123 | 118 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000540739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000540739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000540739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000540739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104172.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104172.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104172.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104172.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104172.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104172.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000540739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000540739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000540739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000540739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104172.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104172.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104172.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104172.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104172.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104172.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104173.1 | Refseq |

LOC283352
Genomic Map