Assay Details
| Gene Symbol: LINC00841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_136148.1 | 2 | 607 | 85 | ||
| GenBank mRNA | AI205837.1 | - | 1 | 85 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00841| Gene Symbol: LINC00841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_136148.1 | 2 | 607 | 85 | ||
| GenBank mRNA | AI205837.1 | - | 1 | 85 | ||
Set Membership: |
|